▸
Histology and Cytology
▸
Tissue Embedding
▸
Embedding Cassettes and Storage
▸
Cassette Storage Cabinet
Actual product may vary
Cassette Storage Cabinet
Economical option for secure storage of tissue cassettes
Have Questions?
Catalog Number
Holds
FIS22-126-345
1500 Tissue
Catalog number
FIS22-126-345
Price (SGD)
-
Holds:
1500 Tissue
Economical option for secure storage of tissue cassettes
Sturdy ABS plastic construction
Six drawers hold up to 1500 standard tissue cassettes
Ergonomic handle for easy opening and closing
Stackable top and bottom design
Overall L x W x H: approx. 17.5 x 9.25 x 15.5 in.
Drawer L x W x H: approx. 15.75 x 9 x 2 in.
Specifications
Holds
1500 Tissue
Depth (English)
17.5 in.
Height (English)
15.5 in.
Width (English)
9.25 in.