Storage File Systems
Actual product may vary
Fisherbrand
Storage File Systems
For long-term storage of histology microscope slide and tissue cassette specimens.
Have Questions?
Catalog NumberdimSlideCapacity
FIS22-126-344
Catalog number FIS22-126-344
Price (SGD)
-
Fisherbrand™ slide and tissue cassette storage boxes provide an economical yet sturdy and secure method for long-term storage of your microscope slide and tissue cassette block specimens.
Our easy to assemble boxes come 10 per case and include 40 drawers/drawer inserts for the slide storage option (4 drawers per file) and 80 drawers/drawer inserts for the tissue block storage option (8 drawers per file).
The coated surface helps to ensure moisture resistance for long-term protection. Each microscope slide storage box holds up to 2000 microscope slides while each tissue block storage box holds up to 500 tissue blocks/cassettes. The unique locking tabs help keep stacked boxes safely secured and stable.
Specifications
ColorWhite
Device Holds500 Tissue Block
For Use With (Application)Histology Speciemen Storage
Height (Metric)100 mm
IncludesOne file box and eight small drawers
MaterialCardboard
Width (Metric)299 mm
Unit SizeCase