Cover Glass Forceps
Cover Glass Forceps
Actual product may vary
Bel-Art
Cover Glass Forceps
Tips shaped to easily lift cover glass. Bel-Art™ Cover Glass Forceps are fully coated with brown Teflon™ FEP and comply with FDA regulations concerning direct food contact.
Have Questions?
Catalog NumberLength (Metric)Tip Style
FIS22-124586102 mmBent
Catalog number FIS22-124586
Price (SGD)
-
Length (Metric):
102 mm
Tip Style:
Bent
  • Brown PTFE FEP coating guards against scratches
  • Wide, bent tip style is perfect for lifting cover glasses
Specifications
Tip StyleBent
Length (English)4 in.
MaterialTeflon
Length (Metric)102 mm
Product TypeForceps
TypeCover Glass Forceps
StyleStandard
Unit SizePack