SP Scienceware™ Plastic Tubing Cutter and Replacement Blades
Actual product may vary
SP Scienceware™ Plastic Tubing Cutter and Replacement Blades
SP Scienceware™ Plastic Tubing Cutter and Replacement Blades
Bel-Art
SP Scienceware™ Plastic Tubing Cutter and Replacement Blades
Portable, pocket-sized cutter cuts flexible tubing from 1.6mm to 19mm (1/16 to 3/4 in.) O.D.
Have Questions?
Catalog NumberProduct Type
FIS22-088245Tubing Cutter
Catalog number FIS22-088245
Price (SGD)
93.00
Each
Product Type:
Tubing Cutter
Price (SGD)
93.00
Each
  • Spring-loaded jaw will remain closed for safety when not in use
  • Replaceable, hardened steel blade makes clean, straight cuts
Not recommended for latex tubing.
Specifications
MaterialPlastic
Overall Length (English)3 in.
Overall Length (Metric)76.2 mm
For Use With1.6 to 19mm Tubing
Product TypeTubing Cutter
Unit SizeEach