Royaltek™ Stainless Steel Surgical Scalpel Blades
Actual product may vary
Royaltek™ Stainless Steel Surgical Scalpel Blades
Surgical Design™ Royaltek™ Stainless Steel Surgical Scalpel Blades are made from sharp, long lasting surgical stainless steel. 10 series blades fit 3, 3L, 3 Graduated, 5B, 7, 9 B3, and B3L scalpel handles. 20 series blades fit 4, 4L, 4 Graduated and 6B.
Have Questions?
Catalog NumberSize
FIS22-079-68310
Catalog number FIS22-079-683
Price (SGD)
157.00
Pack
Size:
10
Price (SGD)
157.00
Pack
  • Surgical Design™ presents Royaltek™- our premium line of surgical blades
  • Exceptionally strong and unifoirmly sharpened to maximum acuity
  • Surgical grade stainless-steel blades
  • Individually sealed
  • Peelable packing
  • Sterile
Specifications
Blade MaterialStainless Steel
Product TypeScalpel Blade
For Use With (Application)Harvesting Radial Arteries during Coronary Artery bypass Operation, Opening the Bronchus during Thoracic Surgery, Inguinal Hernia Repair
Size10
For Use With (Equipment)Fits Handles # 3, # 3L, # 3 Graduated, # 5B, # 7, # 9, # B3 and # B3L.
DescriptionPrimarily used for Small Inscisions in Skin and Muscle