Check Order Status
Spinbar™ Round Magnetic Stirring Bars
Actual product may vary
Bel-Art
Spinbar™ Round Magnetic Stirring Bars
Cylindrical magnetic stirring bars offer excellent centering and smooth running characteristics
Have Questions?
Change view
Catalog Number
Diameter (English)
Diameter (Metric)
Length (English)
Length (Metric)
FIS22-067249
0.31 in.
8 mm
1.75 in.
44.5 mm
FIS14-511-73
0.5 in.
12.7 mm
3 in.
76.2 mm
2 Options
Catalog number
FIS22-067249
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Diameter (English):
0.31 in.
Diameter (Metric):
8 mm
Length (English):
1.75 in.
Length (Metric):
44.5 mm
Small removable pivot ring in center adds to versatility
Pivot ring minimizes contact area of bar to vessel, reduces friction and lessens marring of plastic containers
Alnico magnet encapsulated in FDA Grade, USP Class VI PTFE low friction coating
Durable and inert
Autoclave safe
Specifications
Autoclavable
Autoclavable
Certifications/Compliance
FDA, USP Class VI, ISO 9001:2008
Color
White
Diameter (English)
0.31 in.
Length (English)
1.75 in.
Coating
PTFE, FDA-grade, USP Class VI
Diameter (Metric)
8 mm
Length (Metric)
44.5 mm
Material
Alnico Magnet
Quantity
50
Shape
Cylinder
Type
Magnetic Stir Bar with Pivot Ring