Bransonic™ Ultrasonic Cleaning Bath Accessory, Perforated Trays
Actual product may vary
Branson Ultrasonics
Bransonic™ Ultrasonic Cleaning Bath Accessory, Perforated Trays
For use with Branson ultrasonic baths
Have Questions?
Catalog NumberCapacity (Metric)For Use With (Equipment)
FIS22-06628211.4 LUltrasonic Bath, Model 5800
Catalog number FIS22-066282
Price (SGD)
565.00
Each
Capacity (Metric):
11.4 L
For Use With (Equipment):
Ultrasonic Bath, Model 5800
Price (SGD)
565.00
Each
For use with Branson ultrasonic baths
  • Accessories for ultrasonic bench-top system
Specifications
Product TypePerforated Tray
Capacity (Metric)11.4 L
Capacity (English)2.5 gal.
For Use With (Equipment)Ultrasonic Bath, Model 5800
MaterialStainless Steel
Dimensions (L x W x H)29.2 x 24.1 x 15.2 cm
Unit SizeEach