Glove Box Holder
Glove Box Holder
Actual product may vary
Glove Box Holder
Dispensers ensure that gloves are easily accessible, and help to keep glove boxes from getting crushed or damaged.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacityColor
FIS22-0465593 BoxesGreen
FIS22-0465583 BoxesRed
Catalog number FIS22-046559
Price (SGD)
-
Capacity:
3 Boxes
Color:
Green
  • Designed to help store and organize multiple boxes of disposable gloves
Specifications
Product TypeGlove Box Holder
ColorGreen
Depth (English)4.8 in.
Depth (Metric)12.1 cm
Width (English)11.5 in.
Width (Metric)29.2 cm
Height (English)15.3 in.
Height (Metric)38.8 cm
HoldsDisposable Glove Boxes
MaterialAcrylic
No. of Compartments1
Product StyleTop Loading
Capacity3 Boxes
IncludesTape Tabs, Foam Insert
Unit SizeEA