Check Order Status
Signature Series™ Clear-Rite 3™
Actual product may vary
Thermo Scientific
Signature Series™ Clear-Rite 3™
Render complete deparaffinization and clearing during the staining process with Epredia™ Signature Series Clear-Rite 3.
Have Questions?
Catalog Number
Quantity
FIS22-046341
4 x 1 gal.
Catalog number
FIS22-046341
Price (SGD)
696.00
CS
Add to cart
Quantity:
4 x 1 gal.
Price (SGD)
696.00
CS
Add to cart
Very low odor; Benzene-free
Use in all tissue processors and automatic stainers
Does not produce hard or brittle specimens, even after prolonged exposure
Specifications
Product Type
Clearing Agent
Quantity
4 x 1 gal.