Safety Blood Collection Set
Actual product may vary
Safety Blood Collection Set
Sterile and preassembled for ease of use
Have Questions?
Change viewbuttonViewtableView
Catalog NumberSize
FIS22-040-22423 G
FIS22-040-22321 G
Catalog number FIS22-040-224
Price (SGD)
0.00
Pack
Size:
23 G
Price (SGD)
0.00
Pack
  • Cost effective; ensures your patient only has to undergo one venipuncture for all procedures
  • Allows collection of blood into blood culture bottles and Vacuette™ blood collection tubes in one procedure
  • Safety Blood Collection Set with preattached holder has been designed for use with most commonly used blood culture bottles
  • Compatible with BacT/Alert and BacTec blood culture bottles
  • Standard Blood Collection Set (without safety machanism) is also available with preassembled blood culture holder; contact your Fisher HealthCare Representative
Note: After blood collection, promptly dispose of in an approved sharps disposal container in accordance with the procedures of your facility.
Specifications
Certifications/ComplianceCE, ISO 11014
ColorClear
For Use With (Equipment)Blood culture bottles
Latex FreeYes
Length (English) Tube11.81 in.
Length (Metric) Tube30 cm
Size23 G
Product TypeSafety Blood Collection Set