Wheaton™ BOD Bottles Replacement Robotic Stopper
Wheaton™ BOD Bottles Replacement Robotic Stopper
Actual product may vary
DWK Life Sciences
Wheaton™ BOD Bottles Replacement Robotic Stopper
Compatible with 60 and 300mL BOD bottles
Have Questions?
Change viewbuttonViewtableView
Catalog NumberStopper
FIS22-037004Robotic
FIS02-911-296Pennyhead
Catalog number FIS22-037004
Price (SGD)
217.00
CS
Add to cart
Stopper:
Robotic
Price (SGD)
217.00
CS
Add to cart
  • Glass, closure-style stopper
  • Unlined
Specifications
For Use With (Equipment)For 60 and 300mL BOD bottle
MaterialGlass
StopperRobotic
Unit SizeCS