SuperChip™ Microarray Slides, Epoxysilane
Actual product may vary
Thermo Scientific
SuperChip™ Microarray Slides, Epoxysilane
Choose from several standard surface chemistries and custom coatings with Epredia™ SuperChip™ Microarray Slides.
Have Questions?
Catalog NumberCoatingEnhanced Surface
FIS22-037-216Poly-L-LysineNo
Catalog number FIS22-037-216
Price (SGD)
575.00
Pack
Coating:
Poly-L-Lysine
Enhanced Surface:
No
Price (SGD)
575.00
Pack
Aminopropylsilane-Coated Slides:
  • Ideal for microarrays fabricated with PCR products or proteins as probes
Poly-L-Lysine-Coated Slides:
  • Offer an economical alternative to aminopropylsilane
Aldehyde-Silane-Coated Slides:
  • Provide a uniform coating of aldehydes for the attachment of DNA and protein molecules
Epoxysilane-Coated Slides:
  • Utilize a coating of epoxide groups for the attachment of amine-modified DNA, proteins, peptides, etc. with an amine end
SuperChip Advance:
  • Nitrocellulose-coated slides for use in reverse phase arrays or protein microarray
SmartPlex:
  • Multiplexing microarray platform SmartPlex allows for easy printing of arrays onto glass prior to attaching 96-well upper structure for multiplexing
Product Details:
  • Consistent surface coating to provide a uniform DNA or protein immobilizing surface
  • Optimal amount of probe binded while minimizing nonspecific binding of labeled targets
  • Many available with an enhanced surface (ES) treatment that produces a more uniform, slightly larger spot size without changing surface chemistry
  • Compatible with standard microarray spotting and detection systems
  • Ready for immediate use or may be stored at room temperature for up to a year (six months for Poly-L-Lysine)
  • Printable area: 20 × 70mm
  • Secure no-contact mBox™ packaging, vacuum sealed in a foil barrier bag
Specifications
CoatingPoly-L-Lysine
DescriptionSuperChip Microarray Slides
ColorPlain
MaterialFluorescence Glass
TypeSuperchip Microarray Slide
Length (Metric)70 mm
Dimensions (LxW)70 x 20 mm
Width (Metric)20 mm
Enhanced SurfaceNo
Unit SizePack