Cotton-Tipped Plastic-Shaft Sterile Applicators
Actual product may vary
Puritan
Cotton-Tipped Plastic-Shaft Sterile Applicators
Cotton-tipped sterile applicators with 6 in. plastic shafts
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
FIS22-029-504Individually wrapped
FIS22-029-499Pack of 2
Catalog number FIS22-029-504
Price (SGD)
-
Quantity:
Individually wrapped
  • Semiflexible plastic shaft
  • L x Dia.: 6 x 0.083 in. (15.24 x 0.210cm)
  • Use only cotton grown in the U.S.
  • High-quality, USP-grade cotton fibers are carded and coiled in-house to ensure consistency
Specifications
QuantityIndividually wrapped
Certifications/ComplianceCE, ISO 9001:2008, ISO 13485:2003, FDA GMP's
Diameter (English) Handle0.1 in.
Length (English)6 in.
For Use With (Application)All purpose general swab, wound care, specimen collection
Product TypeApplicator
Diameter (Metric) Handle0.25 cm
Length (Metric)15.2 cm
Length (English) Handle5.87 in.
Length (Metric) Handle1.49 cm
Length (English) Tip0.687 in.
Length (Metric) Tip1.7 cm
Diameter (Metric) Tip0.51 cm
Diameter (English) Tip0.2 in.
SterilitySterile
Handle MaterialPolystyrene
Tip StyleRegular
Tip MaterialCotton
Unit SizeCase