DIFF-SAFE™ Blood Dispenser for Blood Smears
Actual product may vary
Alpha Scientific
DIFF-SAFE™ Blood Dispenser for Blood Smears
Cuts the time of smear-making in half, reducing technician fatigue
Have Questions?
Catalog NumberQuantity
FIS22-026396Case of 10000
Catalog number FIS22-026396
Price (SGD)
597.00
Pack
Quantity:
Case of 10000
Price (SGD)
597.00
Pack
Cuts the time of smear-making in half, reducing technician fatigue
  • Compared to the typical procedure involving 15 or more steps when sticks or capillaries are used, the procedure using the DIFF-SAFE dispenser cuts the number of steps by two thirds
  • Blunt cannula size is 21G x 0.5 in.
Specifications
Product TypeBlood Dispenser
Includes21G x 0.5 in. Blunt needle
For Use With (Application)Blood smears
QuantityCase of 10000
Unit SizePack