▸
Microscope Slides
▸
Glass Microscope Slides
▸
Fisherbrand Formalin Absorbing Pads 275 x 250 mm 6 pcs
Fisherbrand Formalin Absorbing Pads 275 x 250 mm 6 pcs
Fisherbrand Formalin Absorbing Pads 275 x 250 mm 6 pcs
Have Questions?
Catalog number
FIS22-026-70
Price (SGD)
-
Fisherbrand Formalin Absorbing Pads 275 x 250 mm 6 pcs
Specifications
Length (Metric)
275 mm
Width (Metric)
250 mm
Color
Blue