Unistik™ 3 Safety Lancets
Actual product may vary
Unistik™ 3 Safety Lancets
Unistik™ 3 Safety Lancets
Owen Mumford
Unistik™ 3 Safety Lancets
Feature unique Comfort Zone™ technology that allows ultimate control and comfort when lancing
Have Questions?
Catalog NumberColorNeedle GaugeQuantity
FIS22-023-378Yellow23g100 Pack
Catalog number FIS22-023-378
Price (SGD)
-
Color:
Yellow
Needle Gauge:
23g
Quantity:
100 Pack
  • Preset and ready for use; simply twist off protective sterile cap and take sample
  • Needle point is hidden before use and automatically retracts after use to avoid accidental needlestick injuries and cross-infection
  • Have an additional visual lockout feature; should an attempt be made to misuse the device, there is a clear indication to the operator that the device has been used and should be disposed of in a suitable sharps container
Specifications
ColorYellow
Depth (English)0.07 in.
Depth (Metric)1.8mm
DisposableSingle-use
For Use With (Application)For ward blood glucose monitoring, cholesterol, HbA1c
IncludesSterile cap
Needle Gauge23g
Quantity100 Pack
Volume (English)0.00101 to 0.00253 oz.
Volume (Metric)30 to 75μL
Product TypeUnistik™ 3 Safety Lancet
Unit SizePack