Wide Mouth Wash Bottles
Actual product may vary
Bel-Art
Wide Mouth Wash Bottles
Color-coded polypropylene closures for easy identification.
Have Questions?
Catalog NumberClosure Color
FIS22-020253Blue
Catalog number FIS22-020253
Price (SGD)
105.00
Pack
Closure Color:
Blue
Price (SGD)
105.00
Pack
  • Low-density polyethylene (LDPE) wash bottles have wide mouths for easy filling
  • Capacity is 500 mL (16 oz.)
  • Pictogram instructions promote safety regardless of language or setting
Specifications
Capacity16 oz., 500 mL
Capacity (Metric)500 mL
Capacity (English)16 oz.
ColorTranslucent, Blue
MaterialLDPE
SterilityNon-sterile
Closure ColorBlue
Closure IncludedYes
DisposableReusable
GraduatedNo
MouthWide
ShapeCylindrical
Product TypeEconomy Wash Bottle
Chemical LabelUnlabeled