Graduated 15mL Centrifuge Tubes
Actual product may vary
Globe Scientific Inc
Graduated 15mL Centrifuge Tubes
Graduated centrifuge tubes available in polystyrene (PS) or polypropylene (PP) versions.
Have Questions?
Catalog NumberPackagingMaterialClosure Type
FIS22-010-068BulkPolystyrenePlug
Catalog number FIS22-010-068
Price (SGD)
716.00
Case
Packaging:
Bulk
Material:
Polystyrene
Closure Type:
Plug
Price (SGD)
716.00
Case
  • Graduated from 0.5mL to 15mL in 0.5mL increments
  • Conical bottom
  • Polystyrene (PS) spin rate: 1500rcf
  • Polypropylene (PP) spin rate: 3000rcf
  • Dimensions: 16.6 x 120mm
Specifications
Capacity (Metric)15 mL
MaterialPolystyrene
PackagingBulk
SterilityNon-sterile
AutoclavableAutoclavable
Length (Metric)120 mm
Bottom ShapeConical
Closure TypePlug
Diameter (Metric)16 mm
GraduatedYes
Graduations0.5 to 15 mL
Product TypeCentrifuge Tube
Max. RCF1,500 x g
Unit SizeCase