4 Pocket PETG Serological Pipette Holder
Actual product may vary
Poltex
4 Pocket PETG Serological Pipette Holder
The Poltex 4 Pocket Serological Pipette Holder is constructed from durable PETG to provide a lasting, space-saving solution for your serological pipettes.
Have Questions?
Catalog NumberMountingIncludes
FIS22-000-256CountertopPipette Controller Bracket
Catalog number FIS22-000-256
Price (SGD)
759.00
Each
Mounting:
Countertop
Includes:
Pipette Controller Bracket
Price (SGD)
759.00
Each
  • Constructed from durable PETG
  • Holds 4 types of Serological Pipettes
  • Extra storage space on top
  • Shelves are removable for cleaning
  • Choose from powerful, interchangeable magnet mount, adhesive command strip mount, or countertop placement
Specifications
DescriptionWith pipette controller bracket
AutoclavableNot Autoclavable
ColorClear
HoldsSerological Pipets
MaterialPETG
Positions4
For Use With (Application)Serological Pipettes
MountingCountertop
Size4 Pocket
Dimensions (L x W x H)11 x 4.75 x 16 in.
Length (Metric)280 mm
Length (English)11 in.
Width (English)4.6 in.
Height (English)15.9 in.
IncludesPipette Controller Bracket
Product TypeSerological Pipet Holder
Diameter (Metric) Inner28 mm
Height (Metric)405 mm
Width (Metric)116 mm
Unit SizeEach