Three Compartment Benchtop Organizer
Actual product may vary
Three Compartment Benchtop Organizer
Three Compartment Benchtop Organizer
Poltex
Three Compartment Benchtop Organizer
Dispenser helps organize common laboratory items that typically do not have a permanent space on the benchtop. Designed to hold two boxes of disposable gloves, two boxes of lab wipes, with a slide cutter for a roll of parafilm.
Have Questions?
Catalog NumberTotal Positions
FIS22-000-1613
Catalog number FIS22-000-161
Price (SGD)
516.00
Each
Total Positions:
3
Price (SGD)
516.00
Each
  • Constructed from durable, break-resistant 1/4 in. PETG
  • Holds two boxes of disposable gloves, two boxes of lab wipes, and one roll of parafilm
  • Base includes rubber bump-ons to minimize slipping and scratching
  • Consumables are shown for demonstration purposes only
Specifications
Dimensions (D x W x H) Interior6 x 11 x 10.5 in.
HoldsDisposable Glove Boxes, Labwipes, Parafilm
IncludesNon-slick rubber bump-ons
Total Positions3
Product TypeDeluxe Benchtop Organizer
ColorClear
MaterialPETG
Unit SizeEach