Glove Box Holder ST
Actual product may vary
Poltex
Glove Box Holder ST
Holders are constructed of break-resistant PETG and can be mounted horizontally or vertically. Each glove box has its own individual compartment for easy replacement of empty boxes.
Have Questions?
Catalog NumberCapacityMounting
FIS22-000-1393 BoxesMagnetic Mount
Catalog number FIS22-000-139
Price (SGD)
268.00
Each
Capacity:
3 Boxes
Mounting:
Magnetic Mount
Price (SGD)
268.00
Each
  • Constructed of clear, break-resistant PETG
  • Each box has its own individual compartment for easy replacement of empty boxes
  • Holder can be positioned vertically or horizontally
  • Glove boxes are shown for demonstration purposes only
  • Available in magnet mount (powerful magnets included) or wall mount (screws/anchors included)
Specifications
Product TypeGlove Box Holder
ColorClear
Dimensions (L x W x H)9 x 3.95 x 16.5 in.
Length (English)9 in.
Length (Metric)22.9 cm
Width (English)3.95 in.
Width (Metric)10 cm
Height (English)16.5 in.
Height (Metric)41.9 cm
HoldsDisposable Glove Boxes
MaterialPETG
MountingMagnetic Mount
No. of Compartments3
Capacity3 Boxes
IncludesPowerful Magnets
Unit SizeEach