Thermo Scientific
Finnpipette Labtip Blue, 1000/bag
Produced from the highest quality virgin polypropylene
Have Questions?
Catalog NumberVolume (Metric)
FIS21-377-3761000 μL
Catalog number FIS21-377-376
Price (SGD)
139.00
Each
Volume (Metric):
1000 μL
Price (SGD)
139.00
Each
Thermo Scientific™ Labtip standard pipette tips are made from 100% virgin polypropylene. These tips are available in 10, 200, 300 and 1000 μL sizes. Available in clear (10 μL and 300 μL), yellow (200 μL), blue (1000 μL), these tips for general lab use across countless liquid transfer applications.
Labtip pipette tips provide:
  • The most common sizes used in a lab: 10, 200, 300 and 1000 μL
  • Non-Filter essential tips for general laboratory usage
  • Non-sterile packaging in both Racked and Bulk options
Packaging Options:
  • Racked, nonsterile - 96 tips/ tray, 10 trays/pack
  • Bulk - 1,000 tips/ bag
  • Bulk - 25,000 tips/bag
Specifications
Volume (Metric)1000 μL
FilteredNon-filtered
SterilityNon-sterile
ColorBlue
AutoclavableAutoclavable
FormatBulk
Tip StyleUniversal Fit
For Use With (Equipment)Universal Fit
Product LineLabTip