72-Place Wall Mountable Laboratory Drying Racks
72-Place Wall Mountable Laboratory Drying Racks
Actual product may vary
Globe Scientific Inc
72-Place Wall Mountable Laboratory Drying Racks
For draining and drying all types of labware
Have Questions?
Catalog NumberPositions
FIS21-200-29072
Catalog number FIS21-200-290
Price (SGD)
937.00
EA
Add to cart
Positions:
72
Price (SGD)
937.00
EA
Add to cart
  • Closed ended peg sockets to eliminate leakage
  • Seamless drip channel with draining hose
  • Available with 72 large and 11 small interchangeable pegs
  • Supplied with Mounting kit for hanging and hooks for joining additional racks
  • Rust and stain proof
Specifications
MaterialPolystyrene
Positions72
Width (Metric)45 cm
Width (English)17.7 in.
Height (Metric)63 cm
Height (English)24.8 in.
Depth (English)4.3 in.
Depth (Metric)11 cm
Unit SizeEA