Stainless Steel Z-Shaped Racks
Actual product may vary
Globe Scientific Inc
Stainless Steel Z-Shaped Racks
Rust proof racks for repeated use
Have Questions?
Catalog NumberPositionsHolds
FIS21-200-28625Tubes (12 to 13 mm)
Catalog number FIS21-200-286
Price (SGD)
72.00
Each
Positions:
25
Holds:
Tubes (12 to 13 mm)
Price (SGD)
72.00
Each
  • With wide base to provide stability and prevent tipping
  • Stackable inside one another
  • Available as 25-place (5 x 5 format) and 50-Place (5 x 10 format)
Specifications
HoldsTubes (12 to 13 mm)
MaterialStainless Steel
Positions25
Product TypeTube Rack
Unit SizeEach