Product Image
Drummond
BBL™ Dehydrated Culture Media: Thioglycollate Medium Brewer Modified
Dehydrated media for isolation, detection and cultivation of wide variety of microorganisms
Have Questions?
Catalog NumberQuantity
FIS21-171-6500 g
Catalog number FIS21-171-6
Price (SGD)
87.00
Each
Quantity:
500 g
Price (SGD)
87.00
Each
Cultivation of obligate anaerobes, microaerophiles and facultative organism
Specifications
Certifications/ComplianceCE, USP
FormDehydrated
Media TypeThioglycollate Medium Brewer Modified
Product TypeMicrobiology Media
FormulationPancreatic digest of Casein 17.5g/L, Papaic digest of Soybean Meal 2.5g/L, Dextrose 10g/L, Sodium Chloride 5g/L, Sodium Thioglycollate 1g/L, Dipotassium Phosphate 2g/L, Methylene blue 2mg/L, Agar 0.5g/L
For Use With (Application)Cultivation of obligate anaerobes, microaerophiles and facultative organism
Packaging TypeBottle
Quantity500 g
Shelf Life1460 Days
Unit SizeEach
Contents & Storage
2 to 25°C