Check Order Status
Chlorine Water, Saturated, Approximately 0.3% (w/v), Ricca Chemical
Element Search
Structure Search
Chlorine Water, Saturated, Approximately 0.3% (w/v), Ricca Chemical
Have Questions?
Change view
Quantity:
1 L
500 mL
Packaging:
Safety-coated Glass Bottle
Catalog number
FIS1980-32
Price (SGD)
-
Quantity:
1 L
Packaging:
Safety-coated Glass Bottle
Request bulk or custom format
Specifications
Chemical Name or Material
Chlorine Water
Color
Green-Yellow
CAS
7782-50-5
CAS Min %
98.0
CAS Max %
100.0
View more