▸
Buffers and Standards
▸
Matrix Blanks and Standardization Solutions
▸
Chloride Standard, 0.100 N (N/10), 100 mEq/L NaCl, Ricca Chemical
Element Search
Structure Search
Chloride Standard, 0.100 N (N/10), 100 mEq/L NaCl, Ricca Chemical
Have Questions?
Catalog Number
Quantity
Packaging
FIS1960-32
1 L
Natural Poly Bottle
Catalog number
FIS1960-32
Price (SGD)
-
Quantity:
1 L
Packaging:
Natural Poly Bottle
Request bulk or custom format
Specifications
Chemical Name or Material
Chloride Standard
Name Note
100mEq/L NaCl
Packaging
Natural Poly Bottle
CAS
7647-14-5
CAS Min %
97.0
View more