SB 334867
Actual product may vary
SB 334867
SB 334867
Fisherbrand
SB 334867
Selective non-peptide OX1 antagonist
Have Questions?
Catalog NumberQuantity
FIS1960-11 mg
Catalog number FIS1960-1
Price (SGD)
-
Quantity:
1 mg
Selective non-peptide orexin OX1 receptor antagonist. pKb values are 7.2 and < 5 for inhibition of intracellular Ca2+ release in CHO cells expressing human OX1 and OX2 receptors respectively. Blocks orexin-A induced grooming and feeding following systemic administration in vivo.
For Research Use Only
Specifications
Chemical Name or MaterialN-(2-Methyl-6-benzoxazolyl)-N'-1,5-naphthyridin-4-yl urea
CAS792173-99-0
Assay Percent Range>99%
Recommended StorageDesiccate at Room Temperature
Quantity1 mg
SolubilitySoluble to 100mM with gentle warming in DMSO
Molecular FormulaC17H13N5O2
Formula WeightObserved MW: 319.32
Unit SizeEach