Enterprise Services
Vendor Managed Inventory Services
Brands
Shop by Brand
eSolutions
Offers
Webinars
Chemical Structure Search
Support
Fisher Scientific Home
Search
Fisher Scientific
Quick Order
Offers
Sign In
Account options
See All Product Categories
Shop by Brand
Browse All Brands
Services
Enterprise Services
eSolutions
Vendor Managed Inventory Services
Chemical Structure Search
Support
Webinars
Home
▸
Shop All Products
▸
Personal Protective Equipment
▸
Safety Clothing
▸
Aprons
▸
Aprons
Actual product may vary
MCR Safety
Aprons
Protection from oils, chemicals, gritty, grimy work areas and abrasive situations
Have Questions?
Catalog Number
Color
FIS19-822-794
Aqua Green
Catalog number
FIS19-822-794
Price (SGD)
320.00
Dozen
Add to cart
Color:
Aqua Green
Price (SGD)
320.00
Dozen
Add to cart
Heavy-weight PVC construction
Rough edge
One size fits all
Size: 89 × 122cm (35 × 48 in.)
Color: Aqua green
Specifications
Material
PVC
Color
Aqua Green
Dimensions (L x W)
88.9 x 122 cm (35 x 48 in.)
Unit Size
Dozen