Heavy Duty 48-L302 Flock Lined Latex Gloves
Actual product may vary
PIP
Heavy Duty 48-L302 Flock Lined Latex Gloves
Unsupported, heavy duty 28 mil thick latex gloves provide resistance to acids, alkalis, alcohols, salts, ketones and animal fats.
Have Questions?
Catalog NumberSize
FIS19-816-845Large
Catalog number FIS19-816-845
Price (SGD)
-
Size:
Large
For refining, galvanizing, degreasing, plating, petrochemical industries, kitchen cleaning and maintenance
  • Super diamond grip
  • Beaded rolled cuff
  • Thickness: 28 mil
  • Length: 12 in. (30.4 cm)
  • Color: orange
It is strongly advised not to wash and re-use these gloves when they are used to protect against chemical products.
Specifications
Certifications/ComplianceISO 9001:2000, FDA 21 CFR 170-199
ApplicationsIndustrial
ColorOrange
Finger Thickness (English)28 mil
Finger Thickness (Metric)0.71 mm
Latex FreeNo
MaterialLatex
Palm Thickness (English)28 mil
Palm Thickness (Metric)0.71 mm
Product TypeChemical Protection Gloves
SizeLarge
Thickness (English)28 mil
Thickness (Metric)0.71 mm
For Use With (Application)Refining, galvanizing, degreasing, plating, petrochemical industries, kitchen cleaning and maintenance
Glove Cuff StyleBeaded/Rolled
Grip DesignSuper Diamond
Liner MaterialCotton Flock
LiningLined
Model48-L302T/L
Length (English)13 in.
Length (Metric)33 cm
Unit SizePack