G80 Nitrile Chemical Resistant Gloves
Actual product may vary
Ansell
G80 Nitrile Chemical Resistant Gloves
Green, hand-specific gloves provide maximum protection against chemical and mechanical hazards. KleenGuard™ G80 Nitrile Chemical Resistant Gloves include improved, more consistent cotton flock lining for added comfort.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberSize
FIS19-812-4232X-Large
FIS19-812-420Medium
FIS19-812-421Large
Catalog number FIS19-812-423
Price (SGD)
538.00
Case
Size:
2X-Large
Price (SGD)
538.00
Case
  • Hand specific latex-free gloves protect hands against chemical or mechanical hazards
  • Diamond embossed fingertips offer enhanced wet and dry grip
  • Flock-lined for easy donning and extra comfort in extended use
  • Improved AQL of 0.65 versus 1.5, resulting in fewer pinholes
  • Modified Nitrile formulation for more durability and enhanced chemical resistance.
  • Now available in 5 bulk-packed poly bags versus individually wrapped pairs
  • Ideal for: providing PPE category 3 protection, level 4 abrasion resistance and durability in manufacturing, transport, construction, agriculture cleaning and janitorial work, when handling chemicals, oils, grease, acids, caustics or solvents
Specifications
Certifications/ComplianceFDA, ASTM D3767, ASTM D412, ASTM D5151, ASTM D6319
Certifications/Compliance (Europe)CE 0120
ChlorinatedNo
ColorGreen
Finger Thickness (English)15 mil
Finger Thickness (Metric)0.38 mm
Latex FreeYes
MaterialNitrile
Palm Thickness (English)15.7 mil
Palm Thickness (Metric)0.4 mm
Product TypeNitrile Gloves
Size2X-Large
Thickness (English)15 mil
Thickness (Metric)0.38 mm
EN388 StandardEN 388:4101X
EN 421No
EN ISO 374-1Type A
EN ISO 374-1 CHEMICAL LETTER CODESJKLNOP
EN ISO 374 - 5Bacteria, Fungi only
For Use With (Application)Acids, bases, Solvents and Caustics
Grip DesignRough
Liner MaterialCotton Flock
LiningLined
Model94449
SeriesKleenGuard™ G80
Length (English)13 in.
Length (Metric)33 cm
Unit SizeCase