Heavy-Duty Vinyl Apron
Actual product may vary
Heavy-Duty Vinyl Apron
Heavy-Duty Vinyl Apron
PIP
Heavy-Duty Vinyl Apron
Suitable for food handling and preparation, general maintenance and chemical processing
Have Questions?
Catalog NumberMaterial
FIS19-810-610PVC, Vinyl
Catalog number FIS19-810-610
Price (SGD)
282.00
Case
Material:
PVC, Vinyl
Price (SGD)
282.00
Case
  • Heavy-duty 20 mil vinyl apron with neck ties
  • Comfortable to wear with superior flexibility
  • Excellent resistance to punctures and abrasion, chemicals, fats, oils and grease
  • Individually packaged
  • White
Specifications
TypeApron
SizeUniversal
MaterialPVC, Vinyl
ColorWhite
DisposableReusable
Closure TypeTies
SterilityNon-sterile
Thickness20 mil
For Use With (Application)Great for food handling and preparation, industrial maintenance and chemical processing
Certifications/ComplianceFDA 21CFR 170-199
Dimensions (L x W)45 x 35 in. (114.3 x 88.9 cm)
Length (English)45 in.
Length (Metric)114.3 cm
Width (English)35 in.
Width (Metric)88.9 cm
Seam TypeSeamless
Unit SizeCase