Enterprise Services
Vendor Managed Inventory Services
Brands
Shop by Brand
eSolutions
Offers
Webinars
Chemical Structure Search
Support
Fisher Scientific Home
Search
Fisher Scientific
Quick Order
Offers
Sign In
Account options
See All Product Categories
Shop by Brand
Browse All Brands
Services
Enterprise Services
eSolutions
Vendor Managed Inventory Services
Chemical Structure Search
Support
Webinars
Home
▸
Shop All Products
▸
Personal Protective Equipment
▸
Safety Clothing
▸
Aprons
▸
Clear Polyethylene Apron
Actual product may vary
PIP
Clear Polyethylene Apron
Economical protection for light- to medium-duty applications
Have Questions?
Catalog Number
Material
FIS19-810-605
Polyethylene
Catalog number
FIS19-810-605
Price (SGD)
585.00
Case
Add to cart
Material:
Polyethylene
Price (SGD)
585.00
Case
Add to cart
Five layers of protection For use in industrial manufacturing, paper industry, food processing and any heavy dust environment
Specifications
Type
Apron
Size
Universal
Material
Polyethylene
Color
Clear
Disposable
Single-use
Closure Type
Ties
Sterility
Non-sterile
Latex Free
Yes
Thickness
1 mil
Certifications/Compliance
FDA 21CFR 170-199
Dimensions (L x W)
28 x 46 in. (71.1 x 116.8 cm)
Seam Type
Seamless