PolyTack Roll Mop and Accessories
Actual product may vary
Connecticut Clean Room
PolyTack Roll Mop and Accessories
Perfect for cleaning walls, floors, ceiling and hard to reach areas
Have Questions?
Change viewbuttonViewtableView
Catalog NumberMaterialProduct TypeSize
FIS19-808-718Plastic Inner Core FoamTacky Roller45.7 cm (18 in.)
FIS19-161-925MetalExtension Pole122 to 244 cm (48 to 96 in.)
FIS19-003-433Nickel-coated MetalRoller Mop Handle45.7 cm (18 in.)
FIS19-120-2481Plastic Inner CoreTacky Roller45.7 cm (18 in.)
Catalog number FIS19-808-718
Price (SGD)
154.00
Each
Material:
Plastic Inner Core Foam
Product Type:
Tacky Roller
Size:
45.7 cm (18 in.)
Price (SGD)
154.00
Each
  • Helps keep areas free of contamination with easy to use hand or pole attachments
  • Special anti-microbial agents provide long-lasting protection against bacterial growth
  • Plastic inner core insures cleanliness
  • Refills and replacement handles, mops and tacky rollers available
  • 19-823-738 is a film roll only; it does not include  a handle or extension pole
Specifications
Product TypeTacky Roller
Size45.7 cm (18 in.)
For Use With (Application)For Removing Particulate Matter from the Cleanroom
MaterialPlastic Inner Core Foam
Unit SizeEach