BioClean-D™ Disposable Long Sleeve Cover
Actual product may vary
Ansell
BioClean-D™ Disposable Long Sleeve Cover
Extra long length sleeve covers are constructed from antistatic, low-linting CleanTough™ material and are ISO Class 4 compatible.
Have Questions?
Catalog NumberSterility
FIS19-668-340Sterile
Catalog number FIS19-668-340
Price (SGD)
1,271.00
Case
Sterility:
Sterile
Price (SGD)
1,271.00
Case
Extra long length sleeve covers are constructed from antistatic, low-linting CleanTough™ material and are ISO Class 4 compatible.
  • Protective sleeves available in sterile or non-sterile
  • Low linting, anti-static CleanTough™ fabric
  • Elasticated arms and cuffs
  • ISO Class 4 and EU GMP Grade A compatible
  • Universal size
  • Overall length: 500 mm
  • Applications include cleanroom, pharma, controlled environments, 503B facilities/compounding, manufacturing, industrial
Specifications
Product TypeControlled Environment Sleeve
SizeStandard
MaterialDouble Laminate Microporous Fabric
ColorWhite
SterilitySterile
DisposableSingle-use
Closure TypeElastic Ends
ModelBDSC
For Use With (Application)Cleanroom, Clinical, Medical, Laboratory
Certifications/ComplianceASTM D6978-05, ASTM F739-12, ISO 9001:2015
Certifications/Compliance (Europe)(EU)2016/425, EN ISO 13034:2005+A1:2009, EN ISO 14325, EN 1149-5:2008, EN 530, EN 7854, EN 6530, EN 13934-1, EN 9073-4, EN 863, EN 13935-2
Length (English)19.6 in.
Length (Metric)500 mm
Cleanroom ClassClass 10 (ISO 4)
Unit SizeCase