BioClean™ Legion BLA3 Latex Cleanroom Gloves
Actual product may vary
Ansell
BioClean™ Legion BLA3 Latex Cleanroom Gloves
Non-sterile, latex gloves with textured grip offer comfort, flexibility and 16 in. elbow-length protection. Recommended for Class 100/ISO 5 critical environments.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberSize
FIS19-668-203Large
NRTBLA302-MMedium
NRTBLA303-LLarge
Catalog number FIS19-668-203
Price (SGD)
1,848.00
Case
Size:
Large
Price (SGD)
1,848.00
Case
  • AQL 1.5
  • 16 in. (40 cm) Ambidextrous
  • Elbow length for extra protection
  • Textured for improved grip
  • Powder free, beaded cuff
  • Less than 50 μg/gram
  • Latex provides flexibility and comfort
  • Processed to ensure ISO Class 5 Compatibility
  • Complies with the requirements of the Glove Standard EN 420:2003, the European Medical Gloves Standard EN 455 (Parts 1, 2 & 3) & EN 374-1, 2 & 3:2003
  • In accordance with ISO 2859
Specifications
Certifications/Compliance (Europe)CE 0598, EN 420:2003 + A1:2009, EN 421:2010, EN 455 Part 2, EN ISO 374-1:2016, EN ISO 374-5:2016, Category III
Cleanroom ClassClass 100 (ISO 5)
AQL1.5
ColorNatural
DisposableSingle-use
Exterior FinishFully Textured
Finger Thickness (English)7.8 mil
Finger Thickness (Metric)0.20 mm
HandAmbidextrous
Latex FreeNo
MaterialLatex
Palm Thickness (English)6.6 mil
Palm Thickness (Metric)0.17 mm
PowderedPowder Free
Product TypeCleanroom Gloves
SizeLarge
SterilityNon-sterile
Thickness (English)7.8 mil
Thickness (Metric)0.20 mm
Manufacturing/QMS Audit StandardsISO 9001, PPE Regulation 2016 425 Module D
ModelBLA303-L
SeriesBioClean™ Legion BLA3
Length (English)16 in.
Length (Metric)40 cm
Unit SizeCase