BioClean™ Extra BLAS Sterile Latex Cleanroom Gloves
Actual product may vary
Ansell
BioClean™ Extra BLAS Sterile Latex Cleanroom Gloves
Powder free, sterile gloves are flexible and extra long to provide elbow length protection. Recommended for Class 10/ISO 4 Cleanrooms.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberSize
FIS19-668-078Large
FIS19-668-076Small
FKS12735825Small
FIS19-668-077Medium
FKS12745825Medium
FKS12755825Large
Catalog number FIS19-668-078
Price (SGD)
1,286.00
Case
Size:
Large
Price (SGD)
1,286.00
Case
  • Low levels of latex proteins
  • Extended length 16 in. (40 cm)
  • Ambidextrous, beaded cuff
  • Powder free
  • Gamma irradiated
  • Fully textured for improved grip
  • Flexibile and comfortable
  • Easy double-donning
Specifications
Certifications/ComplianceASTM D3578
Certifications/Compliance (Europe)EN 420:2003 + A1:2009 Category III, EN 455 Part 2, EN374-1:20016, EN374-5:2016 and EN421:2010, ISO 11137-1:2006, (EC)1935/2004 and (EC)2023/2006
Cleanroom ClassClass 10 (ISO 4)
AQL1.5
ColorNatural
DisposableSingle-use
Exterior FinishFully Textured
Finger Thickness (English)8.2 mil
Finger Thickness (Metric)0.21 mm
HandAmbidextrous
Laboratory/Cleanroom UseISO Class 4
Latex FreeNo
MaterialLatex
Palm Thickness (English)6.6 mil
Palm Thickness (Metric)0.17 mm
PowderedPowder Free
Product TypeCleanroom Gloves
SizeLarge
SterilitySterile
Thickness (English)8.2 mil
Thickness (Metric)0.21 mm
Manufacturing/QMS Audit StandardsISO 9001, PPE Regulation 2016 425 Module D
ModelBLAS-L
SeriesBioClean™ Extra BLAS
Length (English)16 in.
Length (Metric)40 cm
Unit SizeCase