Enterprise Services
Vendor Managed Inventory Services
Brands
Shop by Brand
eSolutions
Offers
Webinars
Chemical Structure Search
Support
Fisher Scientific Home
Search
Fisher Scientific
Quick Order
Offers
Sign In
Account options
See All Product Categories
Shop by Brand
Browse All Brands
Services
Enterprise Services
eSolutions
Vendor Managed Inventory Services
Chemical Structure Search
Support
Webinars
Home
▸
Shop All Products
▸
Personal Protective Equipment
▸
Safety Clothing
▸
Safety Vests
▸
Self-Extinguishing Safety Vests
Actual product may vary
Fisherbrand
Self-Extinguishing Safety Vests
Pyramex™ Self-Extinguishing Safety Vests with ANSI/ISEA 107-2015 safety compliance
Have Questions?
Catalog Number
Size
FIS19-555-487
Large
Catalog number
FIS19-555-487
Price (SGD)
39.00
Each
Add to cart
Size:
Large
Price (SGD)
39.00
Each
Add to cart
Vest is not fire resistant but it will self-extinguish
Zipper closure
One 2 in. silver reflective stripe applied horizontally across lower chest
Two 2 in. silver reflective stripes run vertically from top of horizontal stripes, over shoulders to top of back side horizontal stripe
Outside left chest pocket measures 5.5 x 6.5 in.
Inside right patch pocket measures 7.5 x 7.5 in.
Specifications
Closure Type
Zipper
Certifications/Compliance
ANSI/ISEA 107-2015, Type R - Class 2
Color
Hi-vis Lime
Size
Large
Material
Polyester Mesh
Size (English) Chest
42 to 44 in.
No. of Pockets
2
Tape Type
China Brand Silver