STERI-BUFFER™ Phosphate Buffer
Actual product may vary
STERI-BUFFER™ Phosphate Buffer
STERI-BUFFER™ Phosphate Buffer
STERI-BUFFER™ Phosphate Buffer
STERI-BUFFER™ Phosphate Buffer
STERI-BUFFER™ Phosphate Buffer for disinfectant preparation and dilution.
Have Questions?
Catalog NumberVolume (Metric)
FIS19-546-92199 mL
Catalog number FIS19-546-921
Price (SGD)
1,457.00
Case
Volume (Metric):
99 mL
Price (SGD)
1,457.00
Case
  • Buffered to a pH of 7.2 ± 0.2
  • Made with USP water for injection filtered at 0.2 μm
  • Filled in easy open-and-close wide-mouth bottles that are scaled on four sides for easy measurement
  • Terminally sterilized through a validated cycle at a SAL level of 10-6
  • Sealed with a no-tamper strip that is broken once the bottle is opened
  • Filled in a ISO 5 cleanroom
  • Validated for sterility
Specifications
Product TypePhosphate Buffer
pH7.2
SterilitySterile
For Use With (Application)For disinfectant preparation and dilution
IncludesCertificate of Analysis, certificate of Sterility
Quantity72 x 99 mL
Shelf Life2 Years
Volume (Metric)99 mL
Unit SizeCase