BioClean-D Long-Length Non-Sterile Hood
Actual product may vary
Fisherbrand
BioClean-D Long-Length Non-Sterile Hood
Non-sterile hood features an extra-long yoke for maximum coverage.
Have Questions?
Catalog NumberMaterial
FIS19-500-863Spunbond Polypropylene
Catalog number FIS19-500-863
Price (SGD)
927.00
Pack
Material:
Spunbond Polypropylene
Price (SGD)
927.00
Pack
  • Lightweight low-linting CleanTough material
  • ESD properties
  • PPE Cat 3 Type PB [6]
  • Silicone free
  • Certifications and Testing:
    • Manufactured in a facility holding ISO 9001:2015 and (EU)2016/425
    • Module D certificationsConforms to Category 3 - Personal Protective Equipment Regulation(EU)2016/425
    • Tested against standard EN ISO 13034:2005+A1:2009 protective clothing against liquid chemicals (Type PB[6])
    • Tested against standard EN ISO 14325 protective clothing against chemicals
    • Tested against standards: EN 1149-5:2008, protective clothing -electrostatic properties; EN 530, Abrasion resistance; EN 7854, Flexcracking resistance; EN 6530, Resistance to penetration by liquids; EN13934-1, Tensile strength; EN 9073-4, Trapezoidal tear resistance; EN863, Puncture resistance; and EN 13935-2, Seam strength.
    • Tested against permeation standard ASTM F739-12
    • Processed in an NEBB certified ISO Class 4 environment
Specifications
Product TypeHood
SizeUniversal
MaterialSpunbond Polypropylene
ColorWhite
Closure TypeElastic
DisposableSingle-use
Design TypeThree-piece
ModelBDHD
Seam TypeBound, Single Needle Stitching
SterilityNon-sterile
Quantity120/Pk.
Hood TypeThree-piece, Elastic face-opening
Unit SizePack