GammaGuard CE™ Sterile Cleanroom Hoods
Actual product may vary
GammaGuard CE™ Sterile Cleanroom Hoods
GammaGuard CE™ Sterile Cleanroom Hoods
International Enviroguard
GammaGuard CE™ Sterile Cleanroom Hoods
GammaGuard CE™ Sterile Cleanroom Hoods are made of a comfortable fabric that protects against various liquids and particulates.
Have Questions?
Catalog NumberQuantity
FIS19-472-641100/Cs.
Catalog number FIS19-472-641
Price (SGD)
961.00
Case
Quantity:
100/Cs.
Price (SGD)
961.00
Case
  • Sterile
  • Each hood is individually packaged and sterilized to a 10-6 sterility assurance level (SAL)
  • Low-linting fabric helps reduce the release of unwanted particulates
  • Designed with tunnelized elastic face providing a more secure closure and stronger seams
  • Hoods include ties in the back to keep strings out of the way
  • GammaGuard CE™ fabric helps keep the wearer cooler
  • Tear-resistant fabric ensures your safety in challenging environments
  • Made in an ISO-certified facility
Specifications
SizeUniversal
MaterialLow Linting Fabric
ColorWhite
Closure TypeTies in Back
Design TypeTunnelized Elastic Face
Latex FreeYes
ModelCE11063CIS
Seam TypeSerged
SterilitySterile
Packaging TypeIndividually Packaged
Quantity100/Cs.
ResistanceLiquids
Unit SizeCase