MicroGuard CE™ Clean Processed Microporous Boot Covers
Actual product may vary
MicroGuard CE™ Clean Processed Microporous Boot Covers
MicroGuard CE™ Clean Processed Microporous Boot Covers
International Enviroguard
MicroGuard CE™ Clean Processed Microporous Boot Covers
Disposable boot covers are made from high MVTR, innovative fabric. Ideal for ISO 5 or higher non-hazardous work environments where reliable liquid and particulate protection is needed.
Have Questions?
Catalog NumberSizeThicknessPackaging Type
FIS19-472-558Large28 milIndividually Packaged
Catalog number FIS19-472-558
Price (SGD)
843.00
Case
Size:
Large
Thickness:
28 mil
Packaging Type:
Individually Packaged
Price (SGD)
843.00
Case
These microporous boot covers are clean processed to offer the perfect balance of safety and comfort. The fabric keeps dust and liquids out while allowing air to flow freely, so your feet stay cool throughout a hard day's work. These disposable boot covers are ideal for non-hazardous work environments, such as controlled environments or cleanrooms, where you need reliable liquid and particulate protection. Boot covers are designed with tunnelized elastic to contain particle shed. Non-skid soles provide added traction to help prevent accidental slips and trips. Each boot cover is individually packaged to help prevent cross-contamination.
  • Non-skid soles help improve traction to prevent accidental slips
  • Elastic closures for snug, secure fit that keeps boot covers in place
  • Available in Bulk or Individually packaged to prevent cross-contamination
Specifications
SizeLarge
MaterialMicroporous Fabric
ColorWhite
Closure TypeElastic Top and Ankles
Sole TypeNon-skid
Seam TypeSerged
Thickness28 mil
Certifications/ComplianceANSI ISEA 101-1996 R2020, ASTM T903, ASTM F1670-07
Packaging Quantity200/Cs.
Packaging TypeIndividually Packaged
Height (English)17 in.
Height (Metric)43.18 cm
Product TypeControlled Environments Boot Covers
Unit SizeCase