Elite Smokers Cease-Fire Cigarette Butt Receptacles Accessories
Actual product may vary
Justrite
Elite Smokers Cease-Fire Cigarette Butt Receptacles Accessories
For safe containment and disposal of cigarette butts
Have Questions?
Catalog NumberProduct TypeQuantity
FIS19-413-833Disposal Bucket Liner10 Pk.
Catalog number FIS19-413-833
Price (SGD)
178.00
Pack
Product Type:
Disposal Bucket Liner
Quantity:
10 Pk.
Price (SGD)
178.00
Pack
  • Anchoring Cable: 91.4cm (36 in.) wire rope cable provides added security or greater stability in severe weather
  • Brass Padlock: 3/16 in. dia. shackle
  • Bucket Liners: special, burn-resistant laminated aluminum/poly construction; include twist-ties; fit any bucket 29.8cm (11.75 in.) wide x 25.4cm (10 in.) high maximum size
See Also:  Justrite™ Elite™ Smokers Cease-Fire™ Cigarette Butt Receptacles (268503).
Specifications
ColorSilver
Dimensions (L x W)18 x 20 in. (45.7 x 50.8 cm)
Length (Metric)45.7 cm
Quantity10 Pk.
Weight (English)2 lb.
Weight (Metric)0.9 kg
Width (Metric)50.8 cm
For Use With (Application)Collecting cigarette butts
For Use With (Equipment)Justrite™ Elite Smokers Cease-Fire Cigarette Butt Receptacles (268503)
Width (English)20 in.
Length (English)18 in.
Product TypeDisposal Bucket Liner
Unit SizePack