▸
Emergency Response Equipment
▸
Fire, Rescue and EMS
▸
Tents and Tarps
▸
Disposable Towel Drape
Actual product may vary
Dynarex
Disposable Towel Drape
For use in minor surgical or therapeutic procedures where a sterile field or drape is required
Have Questions?
Catalog Number
Material
FIS19-310-671
Tissue/Poly/Tissue
Catalog number
FIS19-310-671
Price (SGD)
311.00
Case
Add to cart
Material:
Tissue/Poly/Tissue
Price (SGD)
311.00
Case
Add to cart
Poly-interlined, 3-ply construction (tissue/poly/tissue) provides absorbency while preventing possible moisture or bacteria migration
Drape eliminates all labor costs associated with cloth towels or drapes
Non-fenestrated
Sterile
Measures 45.7 × 66.04cm (18 × 26 in.)
Specifications
Disposable
Single-use
For Use With (Application)
For use in minor surgical or therapeutic procedures
Latex Free
Yes
Sterility
Sterile
Material
Tissue/Poly/Tissue
Product Type
Towel Drape