ESD Safe CleanWIPE™ 1800 Series Cleanroom Swabs
Actual product may vary
ESD Safe CleanWIPE™ 1800 Series Cleanroom Swabs
ESD Safe CleanWIPE™ 1800 Series Cleanroom Swabs
Foamtec
ESD Safe CleanWIPE™ 1800 Series Cleanroom Swabs
Designed to clean static-sensitive assemblies
Have Questions?
Catalog NumberTip StyleShape
FIS19-304-688FlexibleMicro Spear
Catalog number FIS19-304-688
Price (SGD)
204.00
Pack
Tip Style:
Flexible
Shape:
Micro Spear
Price (SGD)
204.00
Pack
Super-soft, open-cell foam is thermally shaped and sealed around ESD safe, solvent-resistant handle to create a precision cleaning swab. Proprietary manufacturing process results in resilient foam cleaning tips that reduce abrasion related particle shedding while enabling efficient cleaning performance. 1800 Series Swabs are used during difficult tasks requiring precision cleaning of lubricant, epoxy, solder ball and solder flux residues.Widely used with IPA in aerospace, medical electronics, optics and disk drive industries
  • Absorbency and abrasion resistance reduces contamination when cleaning sharp or rough surfaces such as suspensions, sliders, HGAs and HSAs
  • Low ionic and NVR Levels
  • Static dissipative swab tip and handle
  • Low tribocharge
  • Silicone and amide free
Specifications
Diameter (English) Handle0.1 in.
Length (English)3 in.
For Use With (Application)Designed to clean static-sensitive assemblies, for removal of contamination from surfaces in critical electronics manufacturing environments
Handle ColorWhite
Product TypeSwab
Diameter (Metric) Handle2.5 mm
Length (Metric)76.2 mm
Length (English) Handle2.71 in.
Length (Metric) Handle68.8 mm
ShapeMicro Spear
SizeMicro
Thickness (Metric)2.5 mm
Length (English) Tip0.41 in.
Length (Metric) Tip10.4 mm
Width (Metric) Tip6.9 mm
Width (English) Tip0.54 in.
Adhesive TypeThermal
Handle MaterialPolypropylene
Tip StyleFlexible
Tip MaterialPolyurethane Foam
Unit SizePack