▸
Personal Protective Equipment
▸
Safety Clothing
▸
Protective Sleeves
▸
Disposable Arm Guards
Actual product may vary
Disposable Arm Guards
Have Questions?
Catalog Number
Material
FIS19-304-142
Polypropylene
Catalog number
FIS19-304-142
Price (SGD)
104.00
Case
Add to cart
Material:
Polypropylene
Price (SGD)
104.00
Case
Add to cart
Specifications
Material
Polypropylene
Color
White
Latex Free
Yes
For Use With (Application)
Protects forearm in many applications
Length (English)
18 in.
Length (Metric)
45.7 cm