Self-Locking Nylon Ties
Actual product may vary
Associated Bag
Self-Locking Nylon Ties
To create tamper-evident seal that must be cut to remove
Have Questions?
Catalog NumberColorTensile Strength (English)Dimensions (L x W)Thickness
FIS19-300-106Black40 lb.8 x 0.13 in. (20.3 x 0.33 cm)0.054 in.
Catalog number FIS19-300-106
Price (SGD)
174.00
Pack
Color:
Black
Tensile Strength (English):
40 lb.
Dimensions (L x W):
8 x 0.13 in. (20.3 x 0.33 cm)
Thickness:
0.054 in.
Price (SGD)
174.00
Pack
  • Tight locking
  • Made of nylon
  • 18.1kg (40 lb.), 22.7kg (50 lb.) tensile strength
  • L x W: 20.3 x 0.33cm (8 in. x 0.13 in.); 27.9 x 0.44cm (11 x 0.17in.)
  • 1.37mm (0.054 in.), 1.34cm (0.053 in.) thickness
  • Available in various colors: Red, black, green , yellow, blue, natural
Applications:
For closing bags, attaching tags, bundling wires, making temporary repairs
Specifications
TypeSelf-locking Tie
MaterialNylon
ColorBlack
Length (Metric)20.3 cm
Length (English)8 in.
Width (Metric)0.33 cm
Width (English)0.13 in.
Dimensions (L x W)8 x 0.13 in. (20.3 x 0.33 cm)
Thickness0.054 in.
Tensile Strength (English)40 lb.
Quantity1000/Pk.
For Use With (Application)Closing bags, attaching tags, bundling wires, making temporary repairs and more
Unit SizePack