Spill Leader Kit™
Actual product may vary
Spill Leader Kit™
Spill Leader Kit™
Spill Leader Kit™
Safetec
Spill Leader Kit™
Multipurpose, wall-mounted, hazardous fluid control kit featuring Red Z™
Have Questions?
Catalog number FIS19-290-175
Price (SGD)
-
Designed for employee protection while handling, cleaning up, or disposing of hazardous liquids. The solid solutions neutralize, stabilize and solidify a variety of aqueous spills.
Specifications
TypeSpill Leader Kit
Includes3 pairs nitrile gloves, red Z™ solidifier (5 oz. bottle), Green Z™ solidifier (5 oz. bottle), Acid Lock™ solidifier (5 oz. bottle), Alky™ solidifier (5oz. bottle), petro lock absorben
Unit SizeEach