Product Image
Tempshield
Cryo-Gloves Small 29 to 32.5 cm Length
Cryo-Gloves Small 29 to 32.5 cm Length
Have Questions?
Catalog number FIS19-276533
Price (SGD)
-
Cryo-Gloves Small 29 to 32.5 cm Length
Specifications
DescriptionCryo-Gloves Blue Wrist Length Cryogenic Gloves
ColorBlue
HandSpecific
SizeSmall
For Use With (Application)Dry Ice Handling, Low/Ultra-Low Temperature Freezers, and Closed Cryogenic Systems
Liner MaterialThermal
SeriesCryo-GlovesTM
Temperature (English) Min-320°F
Temperature (Metric) Min.-196°C
Unit SizePair