Enterprise Services
Vendor Managed Inventory Services
Brands
Shop by Brand
eSolutions
Offers
Webinars
Chemical Structure Search
Support
Fisher Scientific Home
Search
Fisher Scientific
Quick Order
Offers
Sign In
Account options
See all Product Categories
Shop by Brand
Browse All Brands
Services
Enterprise Services
eSolutions
Vendor Managed Inventory Services
Chemical Structure Search
Support
Webinars
Home
▸
Shop All Products
▸
Specialty Lab Glassware
▸
Ampules
▸
SnapIt Ampoule Openers
Actual product may vary
Cincinnati Surgical
SnapIt Ampoule Openers
Safe way to open glass ampoules, avoiding painful and dangerous sharp injuries
Have Questions?
Catalog Number
For Use With (Application)
FIS19-200-247
Opening 1 to 15 mL Ampules
Catalog number
FIS19-200-247
Price (SGD)
685.00
Pack
Add to cart
For Use With (Application):
Opening 1 to 15 mL Ampules
Price (SGD)
685.00
Pack
Add to cart
For use with glass and general ampoules
Available in both Regular (1 to 15mL) and Large (5 to 25mL) sizes
Specifications
Product Type
Ampule Opener
For Use With (Application)
Opening 1 to 15 mL Ampules
For Use With (Equipment)
Glass Ampules